Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017520.3 | NP_059990.2 | 2 | 353 | ||
XM_011535124.2 | XP_011533426.1 | 2 | 354 | |||
GenBank mRNA | AK092556.1 | - | 2 | 313 | ||
AK300258.1 | - | 2 | 353 | |||
AL832864.1 | - | 2 | 302 | |||
BC003542.1 | - | 2 | 261 | 3611719 | ||
DA672302.1 | - | 2 | 353 |