Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004533.3 | NP_004524.3 | 7 | 597 | ||
GenBank mRNA | BC130536.1 | - | 7 | 564 | 40146567 | |
BC136389.1 | - | 7 | 564 | 9020376 | ||
CF553029.1 | - | 7 | 597 | |||
X73113.1 | - | 7 | 565 |