Gene Symbol: NDUFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204055.1 | NP_001190984.1 | 2, 3 | 708 | ||
NM_004549.5 | NP_004540.1 | 2, 3 | 777 | |||
GenBank mRNA | AF070652.1 | - | 2, 3 | 421 | ||
AF087659.1 | - | 2, 3 | 366 | |||
AF087899.1 | - | 2, 3 | 778 | |||
AF369951.1 | - | 2, 3 | 384 | |||
AF369952.1 | - | 2, 3 | 418 | |||
AK022785.1 | - | 2, 3 | 352 | |||
AK023013.1 | - | 2, 3 | 418 | |||
AL050278.1 | - | 2, 3 | 407 | |||
BC007323.2 | - | 2, 3 | 443 | 2990086 | ||
BE740715.1 | - | 2, 3 | 308 | |||
BX281908.1 | - | 2, 3 | 477 | |||
CR533448.1 | - | 2, 3 | 302 |
Gene Symbol: NDUFC2-KCTD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203261.1 | NP_001190190.1 | 2, 3 | 777 | ||
GenBank mRNA | BG723290.1 | - | 2, 3 | 425 |