Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193528.1 | NP_001180457.1 | 4 | 450 | ||
NM_024583.4 | NP_078859.2 | 4 | 541 | |||
XM_005246853.2 | XP_005246910.1 | 4 | 946 | |||
XM_005246854.4 | XP_005246911.1 | 4 | 2106 | |||
XM_005246855.2 | XP_005246912.1 | 4 | 994 | |||
XM_005246856.2 | XP_005246913.1 | 4 | 950 | |||
XM_005246857.2 | XP_005246914.1 | 4 | 650 | |||
XM_011511839.2 | XP_011510141.1 | 3 | 350 | |||
XM_017004909.1 | XP_016860398.1 | 4 | 1538 | |||
XM_017004910.1 | XP_016860399.1 | 4 | 1016 | |||
GenBank mRNA | AK057912.1 | - | 4 | 519 | ||
AK295357.1 | - | 4 | 434 | |||
BC119684.1 | - | 4 | 492 | 40116568 | ||
BC119685.1 | - | 4 | 492 | 40116570 | ||
BM725063.1 | - | 3 | 532 |