Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202406.1 | NP_001189335.1 | 6 | 2479 | ||
NM_001202407.1 | NP_001189336.1 | 6 | 2489 | |||
NM_001202408.1 | NP_001189337.1 | 6 | 2557 | |||
NM_001202409.1 | NP_001189338.1 | 7 | 2779 | |||
NM_001202410.1 | NP_001189339.1 | 7 | 2627 | |||
NM_001202411.1 | NP_001189340.1 | 7 | 2656 | |||
NM_001202412.1 | NP_001189341.1 | 7 | 2342 | |||
NM_032497.2 | NP_115886.1 | 7 | 2542 | |||
GenBank mRNA | AB075488.1 | - | 2 | 1842 | ||
AK027700.1 | - | 7 | 2220 | |||
AK056489.1 | - | 6 | 2054 | |||
AK092348.1 | - | 5 | 2576 | |||
AK298169.1 | - | 4 | 1833 | |||
AL389937.1 | - | 1 | 821 | |||
BC006436.1 | - | 6 | 2527 | 3957973 |