Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138384.2 | NP_612393.2 | 11 | 1210 | ||
GenBank mRNA | AK074976.1 | - | 10 | 1689 | ||
AK095872.1 | - | 5 | 2079 | |||
AK125012.1 | - | 11 | 2055 | |||
AK292515.1 | - | 11 | 1052 | |||
BC000920.2 | - | 9 | 919 | |||
BC004409.1 | - | 11 | 1169 | |||
BC026039.1 | - | 11 | 1186 | 5444745 | ||
BC026725.1 | - | 11 | 1089 | |||
BC035721.1 | - | 11 | 1114 | 5588933 |