Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144661.2 | NP_653262.1 | 5 | 841 | ||
XM_011539344.2 | XP_011537646.1 | 6 | 1235 | |||
XM_011539347.2 | XP_011537649.1 | 5 | 1278 | |||
XM_017015753.1 | XP_016871242.1 | 6 | 856 | |||
XM_017015754.1 | XP_016871243.1 | 6 | 982 | |||
GenBank mRNA | AK097587.1 | - | 5 | 1468 | ||
BC021737.2 | - | 5 | 841 | 4821913 |