Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135556.1 | NP_001129028.1 | 17 | 2767 | ||
NM_001135557.1 | NP_001129029.1 | 16 | 2707 | |||
NM_001278421.1 | NP_001265350.1 | 16 | 2758 | |||
NM_004411.4 | NP_004402.1 | 17 | 2818 | |||
XM_011515861.1 | XP_011514163.1 | 17 | 2665 | |||
XM_011515862.1 | XP_011514164.1 | 17 | 2614 | |||
XM_017011804.1 | XP_016867293.1 | 17 | 3405 | |||
XM_017011805.1 | XP_016867294.1 | 17 | 2592 | |||
GenBank mRNA | AF063228.1 | - | 17 | 2729 | ||
AF123074.1 | - | 16 | 2616 | |||
AK091339.1 | - | 18 | 2667 | |||
BC022540.1 | - | 16 | 2667 | 4793415 | ||
BC032945.2 | - | 17 | 2719 | 4801387 | ||
BC037854.1 | - | 17 | 2709 | 4817657 | ||
BC053542.1 | - | 17 | 2707 | 5727968 |