Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DDIT4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145244.3 | NP_660287.1 | 3 | 377 | ||
GenBank mRNA | AK094236.1 | - | 3 | 347 | ||
AK312926.1 | - | 3 | 347 | |||
AL832277.1 | - | 3 | 329 | |||
BC013592.2 | - | 3 | 335 | 3877854 | ||
DB208869.1 | - | 3 | 377 |