Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300979.1 | NP_001287908.1 | 6 | 2557 | ||
NM_001322911.1 | NP_001309840.1 | 5 | 2430 | |||
NM_001322912.1 | NP_001309841.1 | 5 | 2482 | |||
NM_001322913.1 | NP_001309842.1 | 6 | 2561 | |||
NM_001322914.1 | NP_001309843.1 | 6 | 2733 | |||
NM_001322915.1 | NP_001309844.1 | 7 | 2629 | |||
NM_001322916.1 | NP_001309845.1 | 6 | 2505 | |||
NM_001322917.1 | NP_001309846.1 | 6 | 2509 | |||
NM_001322918.1 | NP_001309847.1 | 7 | 2633 | |||
NM_001322919.1 | NP_001309848.1 | 6 | 2729 | |||
NM_001322920.1 | NP_001309849.1 | 7 | 2681 | |||
NM_152603.4 | NP_689816.2 | 5 | 2434 | |||
XM_011526584.2 | XP_011524886.1 | 4 | 2361 | |||
XM_017026416.1 | XP_016881905.1 | 5 | 2693 | |||
XM_017026417.1 | XP_016881906.1 | 8 | 3776 | |||
XM_017026418.1 | XP_016881907.1 | 6 | 2636 | |||
XM_017026420.1 | XP_016881909.1 | 6 | 3545 | |||
XM_017026421.1 | XP_016881910.1 | 5 | 2656 | |||
GenBank mRNA | AK126691.1 | - | 3 | 3456 | ||
AK303326.1 | - | 6 | 2541 | |||
BC033849.2 | - | 4 | 2408 | 4472579 | ||
BX640706.1 | - | 7 | 2378 |