Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152735.3 | NP_689948.1 | 2 | 1245 | ||
GenBank mRNA | AK122644.1 | - | 2 | 1245 | ||
AK313571.1 | - | 2 | 1279 | |||
BC014978.1 | - | 2 | 1232 | 4874020 |