Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153371.3 | NP_699202.1 | 8 | 1900 | ||
XM_011534995.2 | XP_011533297.1 | 7 | 1531 | |||
XM_017020434.1 | XP_016875923.1 | 8 | 1759 | |||
GenBank mRNA | AK313394.1 | - | 8 | 1900 | ||
BC036755.2 | - | 8 | 1729 | 5541168 |