Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171940.1 | NP_001165411.2 | 6 | 1217 | ||
NM_001171941.2 | NP_001165412.1 | 5 | 2164 | |||
NM_153756.2 | NP_715637.2 | 6 | 1866 | |||
GenBank mRNA | AK092102.1 | - | 6 | 1799 | ||
BX537781.1 | - | 6 | 1093 |