Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148176
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
672
NM_001143806.1
NP_001137278.1
2
887
NM_001143807.1
NP_001137279.1
2
773
NM_001143808.1
NP_001137280.1
2
609
NM_001143809.1
NP_001137281.1
2
619
NM_001143810.1
NP_001137282.1
3
902
NM_001143811.1
NP_001137283.1
4
1019
NM_001143812.1
NP_001137284.1
2
761
NM_001143813.1
NP_001137285.1
2
843
NM_001143814.1
NP_001137286.1
3
968
NM_001143816.1
NP_001137288.1
2
1366
NM_001709.4
NP_001700.2
2
861
NM_170731.4
NP_733927.1
2
1183
NM_170732.4
NP_733928.1
2
970
NM_170733.3
NP_733929.1
2
873
NM_170734.3
NP_733930.1
2
821
NM_170735.5
NP_733931.1
1
1600
XM_011520280.2
XP_011518582.1
2
1767
GenBank mRNA
AF400438.1
-
1
533
AK289763.1
-
2
669
AK289853.1
-
2
570
AY054392.2
-
2
874
AY054393.2
-
2
616
AY054394.2
-
2
831
AY054395.2
-
2
914
AY054396.2
-
2
672
AY054397.2
-
2
755
AY054398.2
-
2
802
AY054399.2
-
3
687
AY054400.2
-
2
788
AY656701.1
-
1
515
BC029795.1
-
2
846
5193877
EF674517.1
-
2
672
EF674518.1
-
2
887
EF674519.1
-
2
970
EF674520.1
-
2
773
EF674521.1
-
2
873
EF689009.1
-
1
1614
EF689010.1
-
2
1380
EF689011.1
-
2
609
EF689012.1
-
2
619
EF689013.1
-
2
761
EF689014.1
-
2
843
EF689015.1
-
2
861
EF689016.1
-
3
968
EF689017.1
-
3
720
EF689018.1
-
2
821
EF689019.1
-
3
902
EF689020.1
-
4
1019
EF689021.1
-
2
1183
KC855559.1
-
1
51
KP229412.1
-
1
515
KU177943.1
-
1
515
M61176.1
-
1
1077
X60201.1
-
2
811
X91251.1
-
2
799
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: AY176065.
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0