Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB2 SIRNA Assays ›
Gene Symbol:
APBB2
Gene Name:
amyloid beta precursor protein binding family B member 2
Gene Aliases:
FE65L, FE65L1
Chromosome Location:
Chr.4: 40810027 - 41216714 on Build GRCh38
UniGene ID:
Hs.479602
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148181
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166050.1
NP_001159522.1
18
3305
NM_001166051.1
NP_001159523.1
8
1326
NM_001166052.1
NP_001159524.1
7
1320
NM_001166053.1
NP_001159525.1
8
1322
NM_001166054.1
NP_001159526.1
7
1316
NM_004307.1
NP_004298.1
18
3308
NM_173075.4
NP_775098.2
17
3239
XM_005248101.3
XP_005248158.1
17
3239
XM_006714005.3
XP_006714068.1
19
3311
XM_006714007.3
XP_006714070.1
19
3309
XM_006714008.3
XP_006714071.1
18
3213
XM_006714009.3
XP_006714072.1
18
3211
XM_006714010.3
XP_006714073.1
19
3308
XM_006714011.3
XP_006714074.1
18
3248
XM_006714012.3
XP_006714075.1
18
3245
XM_011513686.2
XP_011511988.1
20
3333
XM_011513687.2
XP_011511989.1
17
3242
XM_017008142.1
XP_016863631.1
19
3232
XM_017008143.1
XP_016863632.1
20
3330
XM_017008144.1
XP_016863633.1
18
3210
XM_017008145.1
XP_016863634.1
17
3207
XM_017008146.1
XP_016863635.1
18
3245
XM_017008147.1
XP_016863636.1
18
3240
XM_017008148.1
XP_016863637.1
17
3144
XM_017008149.1
XP_016863638.1
18
3242
XM_017008150.1
XP_016863639.1
18
3169
XM_017008151.1
XP_016863640.1
17
3150
XM_017008152.1
XP_016863641.1
17
3147
XM_017008153.1
XP_016863642.1
17
3163
XM_017008154.1
XP_016863643.1
16
3144
XM_017008155.1
XP_016863644.1
16
3141
XM_017008156.1
XP_016863645.1
17
3182
XM_017008157.1
XP_016863646.1
16
3081
XM_017008158.1
XP_016863647.1
16
3176
GenBank mRNA
AK226179.1
-
18
3222
AK299800.1
-
7
1291
AK304247.1
-
18
3176
BC027946.1
-
17
3079
5203925
BC088372.1
-
9
1496
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
regulation of transcription, DNA-templated
cell cycle arrest
axon guidance
extracellular matrix organization
negative regulation of cell growth
intracellular signal transduction
positive regulation of apoptotic process
negative regulation of apoptotic process
beta-amyloid binding
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline