Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173582.4 | NP_775853.2 | 3, 4 | 692 | ||
XM_011544953.2 | XP_011543255.1 | 4, 5 | 747 | |||
GenBank mRNA | AB019210.1 | - | 3, 4 | 643 | ||
AK056591.1 | - | 3, 4 | 650 | |||
BC059360.1 | - | 3, 4 | 656 | 30332467 |