Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173582.4 | NP_775853.2 | 14 | 4195 | ||
XM_011544953.2 | XP_011543255.1 | 15 | 4250 | |||
GenBank mRNA | AB019210.1 | - | 14 | 4145 | ||
AL119265.1 | - | 1 | 267 |