Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173825.3 | NP_776186.2 | 8 | 915 | ||
XM_006713595.2 | XP_006713658.1 | 7 | 827 | |||
XM_017006227.1 | XP_016861716.1 | 9 | 941 | |||
XM_017006228.1 | XP_016861717.1 | 8 | 869 | |||
GenBank mRNA | AK025772.1 | - | 5 | 611 | ||
AK293085.1 | - | 8 | 899 | |||
BC020832.1 | - | 8 | 894 | 4771781 | ||
BX647559.1 | - | 8 | 912 | |||
BX647593.1 | - | 8 | 915 | |||
BX647664.1 | - | 8 | 899 |