Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317031.1 | NP_001303960.1 | 2 | 238 | ||
NM_001317032.1 | NP_001303961.1 | 2 | 238 | |||
NM_001317033.1 | NP_001303962.1 | 2 | 351 | |||
NM_182513.3 | NP_872319.1 | 2 | 238 | |||
XM_005259753.3 | XP_005259810.1 | 2 | 423 | |||
XM_011527702.1 | XP_011526004.1 | 2 | 208 | |||
GenBank mRNA | AK075287.1 | - | 2 | 208 | ||
AK303157.1 | - | 2 | 233 | |||
AK303266.1 | - | 2 | 346 | |||
AY456387.1 | - | 2 | 202 | |||
BC105037.1 | - | 2 | 234 | 8144040 | ||
BC105039.1 | - | 2 | 234 | 8144042 | ||
BQ061822.1 | - | 2 | 195 | |||
BQ216591.1 | - | 2 | 214 | |||
HY082333.1 | - | 2 | 241 |