Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRK4 SIRNA Assays ›
Gene Symbol:
GRK4
Gene Name:
G protein-coupled receptor kinase 4
Gene Aliases:
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location:
Chr.4: 2963504 - 3040760 on Build GRCh38
UniGene ID:
Hs.32959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148245
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004056.1
NP_001004056.1
5, 6
903
NM_001004057.1
NP_001004057.1
6, 7
999
NM_005307.2
NP_005298.2
5, 6
903
NM_182982.2
NP_892027.2
6, 7
999
XM_005247962.3
XP_005248019.1
4, 5
517
XM_006713880.3
XP_006713943.1
5, 6
415
XM_011513447.2
XP_011511749.1
6, 7
1111
XM_011513448.2
XP_011511750.1
6, 7
1111
XM_011513449.2
XP_011511751.1
5, 6
1015
XM_011513450.2
XP_011511752.1
6, 7
1111
XM_011513451.2
XP_011511753.1
6, 7
1111
XM_011513452.2
XP_011511754.1
6, 7
1111
XM_011513453.2
XP_011511755.1
6, 7
1111
XM_011513454.2
XP_011511756.1
6, 7
1111
XM_011513455.2
XP_011511757.1
6, 7
1111
XM_011513456.2
XP_011511758.1
6, 7
1111
XM_011513457.2
XP_011511759.1
6, 7
1111
XM_017008052.1
XP_016863541.1
5, 6
1033
XM_017008053.1
XP_016863542.1
6, 7
1111
XM_017008054.1
XP_016863543.1
6, 7
1111
XM_017008055.1
XP_016863544.1
5, 6
1015
XM_017008056.1
XP_016863545.1
6, 7
1111
XM_017008061.1
XP_016863550.1
5, 6
766
XM_017008062.1
XP_016863551.1
5, 6
913
XM_017008065.1
XP_016863554.1
6, 7
1111
GenBank mRNA
AK223581.1
-
5, 6
911
AK292390.1
-
6, 7
1044
BC117320.1
-
6, 7
887
40125871
U33054.1
-
6, 7
798
U33055.1
-
5, 6
702
U33056.1
-
6, 7
798
X97879.1
-
5, 6
546
X97880.1
-
5, 6
546
X97881.1
-
6, 7
642
X98118.1
-
6, 7
642
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor internalization
protein phosphorylation
signal transduction
regulation of G-protein coupled receptor protein signaling pathway
regulation of rhodopsin mediated signaling pathway
receptor internalization
G-protein coupled receptor kinase activity
ATP binding
rhodopsin kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0