Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005436.4 | NP_005427.2 | 7 | 1328 | ||
GenBank mRNA | AK056848.1 | - | 1 | 1148 | ||
AK292593.1 | - | 7 | 1328 | |||
BC036757.2 | - | 7 | 1167 | 5549623 | ||
BC064391.1 | - | 7 | 1154 | 5745698 | ||
S72869.1 | - | 7 | 1132 |