Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014109.3 | NP_054828.2 | 2 | 323 | ||
XM_011516994.2 | XP_011515296.1 | 2 | 850 | |||
XM_011516995.2 | XP_011515297.1 | 2 | 850 | |||
GenBank mRNA | AK097133.1 | - | 2 | 305 | ||
AL833653.1 | - | 2 | 77 | |||
AY598335.1 | - | 2 | 215 | |||
BC113656.1 | - | 2 | 237 | 8322708 | ||
BP314109.1 | - | 2 | 323 | |||
CR749832.1 | - | 3 | 580 |