Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014824.2 | NP_055639.2 | 8 | 839 | ||
XM_011545409.1 | XP_011543711.1 | 8 | 721 | |||
XM_011545410.2 | XP_011543712.1 | 8 | 654 | |||
XM_017018632.1 | XP_016874121.1 | 8 | 898 | |||
GenBank mRNA | AB018312.1 | - | 7 | 689 | ||
AK296376.1 | - | 7 | 509 | |||
AK297928.1 | - | 8 | 723 | |||
AL133567.1 | - | 3 | 207 | |||
BC010394.1 | - | 8 | 586 | 4091973 |