Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 48, 49 | 5746 | ||
XM_005247245.4 | XP_005247302.1 | 49, 50 | 5943 | |||
XM_017006036.1 | XP_016861525.1 | 35, 36 | 4203 | |||
GenBank mRNA | AB014578.2 | - | 48, 49 | 5795 | ||
AK023168.1 | - | 17, 18 | 2174 | |||
AK125330.1 | - | 25, 26 | 3130 | |||
AK127112.1 | - | 32, 33 | 3834 | |||
AY779857.1 | - | 48, 49 | 5746 | |||
BC000164.3 | - | 1, 2 | 109 | |||
BC009630.1 | - | 3, 4 | 229 | |||
BC151246.1 | - | 49, 50 | 5816 | 9007477 |