Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INTU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015693.3 | NP_056508.2 | 1, 2 | 241 | ||
XM_011531844.2 | XP_011530146.1 | 1, 2 | 268 | |||
XM_011531845.2 | XP_011530147.1 | 1, 2 | 268 | |||
XM_011531849.2 | XP_011530151.1 | 1, 2 | 268 | |||
XM_011531850.2 | XP_011530152.1 | 1, 2 | 268 | |||
XM_011531851.2 | XP_011530153.1 | 1, 2 | 268 | |||
XM_017008026.1 | XP_016863515.1 | 1, 2 | 268 | |||
GenBank mRNA | AB033110.1 | - | 1, 2 | 208 | ||
AK291481.1 | - | 1, 2 | 223 | |||
BC051698.1 | - | 1, 2 | 216 | |||
BC130611.1 | - | 1, 2 | 207 | 40146642 | ||
DB195553.1 | - | 1, 2 | 241 |