Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFTPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002243.2 | NP_001002243.1 | 2 | 2124 | ||
NM_017657.4 | NP_060127.3 | 2 | 2124 | |||
NM_203437.3 | NP_982261.2 | 2 | 2124 | |||
XM_005264380.1 | XP_005264437.1 | 2 | 2124 | |||
GenBank mRNA | AB073356.1 | - | 1 | 1566 | ||
AK000087.1 | - | 2 | 2099 | |||
AK021899.1 | - | 1 | 747 | |||
AK222821.1 | - | 2 | 2047 | |||
AL833962.2 | - | 2 | 1902 | |||
AY367088.1 | - | 1 | 1784 | |||
BC047529.1 | - | 2 | 2098 | 5311682 |