Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFTPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002243.2 | NP_001002243.1 | 9 | 3139 | ||
NM_017657.4 | NP_060127.3 | 9 | 3142 | |||
NM_203437.3 | NP_982261.2 | 10 | 3223 | |||
XM_005264380.1 | XP_005264437.1 | 10 | 3226 | |||
GenBank mRNA | AK000087.1 | - | 9 | 3117 | ||
AK024658.1 | - | 7 | 778 | |||
AK222821.1 | - | 9 | 3065 | |||
AL833962.2 | - | 9 | 4647 | |||
BC022247.1 | - | 6 | 630 | |||
BC047529.1 | - | 9 | 3116 | 5311682 |