Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020964.2 | NP_066015.2 | 28 | 5008 | ||
XM_011526122.1 | XP_011524424.1 | 28 | 5008 | |||
XM_017025889.1 | XP_016881378.1 | 28 | 4990 | |||
XM_017025890.1 | XP_016881379.1 | 28 | 4990 | |||
XM_017025891.1 | XP_016881380.1 | 27 | 4867 | |||
XM_017025892.1 | XP_016881381.1 | 27 | 5033 | |||
XM_017025893.1 | XP_016881382.1 | 20 | 2880 | |||
GenBank mRNA | AK023817.1 | - | 12 | 1687 | ||
BC130614.1 | - | 11 | 1541 | |||
BC144104.1 | - | 11 | 1541 | |||
BC144105.1 | - | 11 | 1541 |