Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022491.2 | NP_071936.2 | 7, 8 | 760 | ||
XM_011538671.2 | XP_011536973.1 | 8, 9 | 1837 | |||
XM_017019817.1 | XP_016875306.1 | 6, 7 | 644 | |||
XM_017019818.1 | XP_016875307.1 | 9, 10 | 992 | |||
XM_017019819.1 | XP_016875308.1 | 8, 9 | 854 | |||
XM_017019820.1 | XP_016875309.1 | 9, 10 | 886 | |||
GenBank mRNA | AK023801.1 | - | 2, 3 | 236 | ||
AK024460.1 | - | 7, 8 | 743 | |||
BC030252.1 | - | 1, 2 | 133 | |||
BC093990.1 | - | 2, 3 | 190 | 7939835 | ||
BC098404.1 | - | 7, 8 | 783 | 30332266 | ||
BC112000.1 | - | 2, 3 | 190 | |||
BX647687.1 | - | 8, 9 | 718 |