Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318099.1 | NP_001305028.1 | 5 | 2302 | ||
NM_001318100.1 | NP_001305029.1 | 5 | 2379 | |||
NM_001318101.1 | NP_001305030.1 | 3 | 1580 | |||
NM_032429.3 | NP_115805.1 | 5 | 2409 | |||
XM_005270222.4 | XP_005270279.1 | 5 | 2641 | |||
XM_017016780.1 | XP_016872269.1 | 4 | 4770 | |||
XM_017016781.1 | XP_016872270.1 | 4 | 1750 | |||
GenBank mRNA | AL834338.1 | - | 3 | 1785 | ||
BC005855.2 | - | 2 | 1113 | |||
BC006212.2 | - | 5 | 2267 | 3161855 |