Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032689.4 | NP_116078.4 | 3, 4 | 743 | ||
XM_006723435.3 | XP_006723498.1 | 3, 4 | 997 | |||
GenBank mRNA | AK131557.1 | - | 3, 4 | 743 |