Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144034.1 | NP_001137506.1 | 8 | 1209 | ||
NM_001144035.1 | NP_001137507.1 | 8 | 1206 | |||
NM_001144036.1 | NP_001137508.1 | 7 | 897 | |||
NM_001318757.1 | NP_001305686.1 | 8 | 1050 | |||
NM_032780.3 | NP_116169.2 | 9 | 1341 | |||
NR_134852.1 | 6 | 800 | ||||
XM_011543037.2 | XP_011541339.1 | 9 | 1222 | |||
XM_017018422.1 | XP_016873911.1 | 9 | 1250 | |||
XM_017018423.1 | XP_016873912.1 | 9 | 1247 | |||
XM_017018427.1 | XP_016873916.1 | 9 | 1131 | |||
XM_017018428.1 | XP_016873917.1 | 9 | 1154 | |||
XM_017018429.1 | XP_016873918.1 | 9 | 1128 | |||
XM_017018430.1 | XP_016873919.1 | 8 | 1090 | |||
XM_017018432.1 | XP_016873921.1 | 8 | 999 | |||
XM_017018433.1 | XP_016873922.1 | 8 | 1022 | |||
GenBank mRNA | AK056325.1 | - | 7 | 783 | ||
AK096260.1 | - | 6 | 1910 | |||
AK124814.1 | - | 6 | 2059 | |||
AY358919.1 | - | 8 | 1094 | |||
BC036433.1 | - | 2 | 195 | |||
BC042896.1 | - | 9 | 1220 | 5168151 | ||
BC051841.1 | - | 7 | 1077 | 5725777 |