Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM25 SIRNA Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.11: 118531088 - 118546670 on Build GRCh38
UniGene ID:
Hs.564188
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
149009
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144034.1
NP_001137506.1
8
2261
NM_001144035.1
NP_001137507.1
8
2258
NM_001144036.1
NP_001137508.1
7
1949
NM_001318755.1
NP_001305684.1
9
1389
NM_001318757.1
NP_001305686.1
8
2102
NM_032780.3
NP_116169.2
9
2393
NR_134852.1
6
1852
XM_005271703.1
XP_005271760.1
9
1301
XM_006718926.2
XP_006718989.1
9
1276
XM_006718927.2
XP_006718990.1
8
1144
XM_011543037.2
XP_011541339.1
9
2274
XM_011543038.2
XP_011541340.1
8
1141
XM_017018422.1
XP_016873911.1
9
2302
XM_017018423.1
XP_016873912.1
9
2299
XM_017018424.1
XP_016873913.1
9
1182
XM_017018425.1
XP_016873914.1
9
1205
XM_017018426.1
XP_016873915.1
8
1169
XM_017018427.1
XP_016873916.1
9
2183
XM_017018428.1
XP_016873917.1
9
2206
XM_017018429.1
XP_016873918.1
9
2180
XM_017018430.1
XP_016873919.1
8
2142
XM_017018432.1
XP_016873921.1
8
2051
XM_017018433.1
XP_016873922.1
8
2074
GenBank mRNA
AK056325.1
-
7
1835
AK096260.1
-
6
2962
AK124814.1
-
6
3111
AK225415.1
-
9
1275
AY358919.1
-
8
2146
BC036433.1
-
2
1247
BC042896.1
-
9
2264
5168151
BC051841.1
-
7
2121
5725777
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-nglqg:80/10.239.166.193:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0