Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308089.1 | NP_001295018.1 | 4, 5 | 592 | ||
NM_001324130.1 | NP_001311059.1 | 4, 5 | 800 | |||
NM_133450.3 | NP_597707.1 | 5, 6 | 794 | |||
NR_040252.1 | 4, 5 | 797 | ||||
XM_005255119.3 | XP_005255176.1 | 3, 4 | 494 | |||
XM_011522372.1 | XP_011520674.1 | 4, 5 | 724 | |||
XM_011522373.1 | XP_011520675.1 | 4, 5 | 606 | |||
XM_011522378.1 | XP_011520680.1 | 3, 4 | 351 | |||
GenBank mRNA | AB075857.1 | - | 2, 3 | 1230 | ||
AK057329.1 | - | 2, 3 | 395 | |||
AK123496.1 | - | 5, 6 | 795 | |||
AK301683.1 | - | 3, 4 | 422 | |||
BC050387.1 | - | 4, 5 | 433 | 5751728 | ||
BM545219.1 | - | 4, 5 | 643 | |||
DR004376.1 | - | 3, 4 | 530 |