Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242929.1 | NP_001229858.1 | 9, 10 | 1306 | ||
NM_001308089.1 | NP_001295018.1 | 11, 12 | 1496 | |||
NM_001324129.1 | NP_001311058.1 | 8, 9 | 1134 | |||
NM_001324130.1 | NP_001311059.1 | 11, 12 | 1699 | |||
NM_133450.3 | NP_597707.1 | 12, 13 | 1698 | |||
NR_040252.1 | 11, 12 | 1701 | ||||
XM_005255119.3 | XP_005255176.1 | 10, 11 | 1398 | |||
XM_005255121.3 | XP_005255178.1 | 8, 9 | 1096 | |||
XM_011522372.1 | XP_011520674.1 | 11, 12 | 1628 | |||
XM_011522373.1 | XP_011520675.1 | 11, 12 | 1510 | |||
XM_011522374.1 | XP_011520676.1 | 10, 11 | 1244 | |||
XM_011522378.1 | XP_011520680.1 | 11, 12 | 1366 | |||
XM_017022935.1 | XP_016878424.1 | 9, 10 | 1306 | |||
XM_017022936.1 | XP_016878425.1 | 11, 12 | 1619 | |||
GenBank mRNA | AB075857.1 | - | 9, 10 | 2134 | ||
AK056907.1 | - | 1, 2 | 1247 | |||
AK057329.1 | - | 8, 9 | 1857 | |||
AK123496.1 | - | 12, 13 | 1699 | |||
AK301683.1 | - | 10, 11 | 1326 | |||
BC050387.1 | - | 11, 12 | 1337 | 5751728 |