Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFYVE27 SIRNA Assays ›
Gene Symbol:
ZFYVE27
Gene Name:
zinc finger FYVE-type containing 27
Gene Aliases:
PROTRUDIN, SPG33
Chromosome Location:
Chr.10: 97737121 - 97760907 on Build GRCh38
UniGene ID:
Hs.744075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
149130
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFYVE27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002261.3
NP_001002261.1
5
626
NM_001002262.3
NP_001002262.1
6
831
NM_001174119.1
NP_001167590.1
5
731
NM_001174120.1
NP_001167591.1
4
569
NM_001174121.1
NP_001167592.1
4
558
NM_001174122.1
NP_001167593.1
3
473
NM_144588.6
NP_653189.3
6
831
XM_005269502.3
XP_005269559.1
6
820
XM_005269503.3
XP_005269560.1
6
824
XM_005269504.3
XP_005269561.1
6
820
XM_005269505.4
XP_005269562.1
6
820
XM_005269506.3
XP_005269563.1
6
820
XM_005269508.3
XP_005269565.1
5
749
XM_005269509.3
XP_005269566.1
5
753
XM_005269510.3
XP_005269567.1
4
565
XM_005269511.3
XP_005269568.1
4
571
XM_011539252.2
XP_011537554.1
6
820
XM_011539253.2
XP_011537555.1
6
820
XM_011539254.2
XP_011537556.1
6
795
XM_011539255.2
XP_011537557.1
6
802
XM_017015644.1
XP_016871133.1
6
820
XM_017015645.1
XP_016871134.1
6
820
XM_017015646.1
XP_016871135.1
6
820
XM_017015647.1
XP_016871136.1
6
824
XM_017015648.1
XP_016871137.1
4
565
XM_017015649.1
XP_016871138.1
4
565
XM_017015650.1
XP_016871139.1
6
795
XM_017015651.1
XP_016871140.1
5
749
XM_017015652.1
XP_016871141.1
5
753
XM_017015653.1
XP_016871142.1
5
749
XM_017015654.1
XP_016871143.1
5
753
XM_017015655.1
XP_016871144.1
4
571
GenBank mRNA
AK057481.1
-
6
802
AK074876.1
-
6
812
AK097945.1
-
4
562
AK296295.1
-
4
558
AK296588.1
-
5
676
AK299735.1
-
3
466
AK300438.1
-
6
824
AL561314.3
-
6
743
AL834235.1
-
5
560
BC030621.2
-
6
824
4818199
BI758753.1
-
6
820
BU838354.1
-
2
149
BX647836.1
-
6
766
KU178847.1
-
4
421
KU178848.1
-
4
421
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
neuron projection development
neurotrophin TRK receptor signaling pathway
protein localization to plasma membrane
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0