Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127361.2 | NP_001120833.1 | 9 | 2181 | ||
NM_001300826.1 | NP_001287755.1 | 9 | 2131 | |||
NM_153341.3 | NP_699172.2 | 9 | 2134 | |||
XM_006710356.2 | XP_006710419.1 | 9 | 2587 | |||
XM_006710357.3 | XP_006710420.1 | 8 | 2455 | |||
XM_006710358.3 | XP_006710421.1 | 9 | 2637 | |||
GenBank mRNA | AK074486.1 | - | 9 | 1434 | ||
AK131439.1 | - | 10 | 1878 | |||
BC020595.2 | - | 5 | 839 | |||
BC062374.1 | - | 9 | 1658 | |||
BC113560.1 | - | 9 | 1501 | 8322612 | ||
BC143687.1 | - | 9 | 1498 | 9052201 |