Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178563.3 | NP_848658.3 | 10 | 1900 | ||
XM_017012136.1 | XP_016867625.1 | 10 | 3222 | |||
XM_017012137.1 | XP_016867626.1 | 10 | 3504 | |||
XM_017012138.1 | XP_016867627.1 | 9 | 1706 | |||
XM_017012139.1 | XP_016867628.1 | 9 | 1706 | |||
XM_017012140.1 | XP_016867629.1 | 9 | 1706 | |||
XM_017012141.1 | XP_016867630.1 | 9 | 3039 | |||
XM_017012142.1 | XP_016867631.1 | 9 | 1706 | |||
XM_017012143.1 | XP_016867632.1 | 9 | 1706 | |||
GenBank mRNA | AK302827.1 | - | 10 | 1900 | ||
BC030651.2 | - | 10 | 1875 | 5267615 |