Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043318.2 | NP_001036783.1 | 3 | 1954 | ||
NM_194278.3 | NP_919254.2 | 3 | 2333 | |||
XM_005268204.1 | XP_005268261.1 | 3 | 2333 | |||
XM_005268205.1 | XP_005268262.1 | 3 | 1889 | |||
XM_005268206.1 | XP_005268263.1 | 4 | 2106 | |||
GenBank mRNA | AK127785.1 | - | 3 | 2332 | ||
BC003121.1 | - | 2 | 1016 | |||
BC006451.1 | - | 2 | 788 | |||
BC006511.2 | - | 2 | 1584 | |||
BC009202.2 | - | 3 | 1954 | |||
BC015668.2 | - | 3 | 1967 | |||
BC025330.1 | - | 2 | 1584 |