Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004661.3 | NP_004652.2 | 2, 3 | 273 | ||
GenBank mRNA | AB011472.1 | - | 2, 3 | 260 | ||
AF053977.1 | - | 2, 3 | 240 | |||
AF191341.1 | - | 2, 3 | 224 | |||
AK291610.1 | - | 2, 3 | 255 | |||
BC005258.1 | - | 2, 3 | 252 | |||
BC010944.1 | - | 2, 3 | 257 | |||
BC017713.1 | - | 2, 3 | 232 | 4399056 | ||
BT009810.1 | - | 2, 3 | 224 |