Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107987389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030147.1 | XP_016885636.1 | 1 | 477 | ||
XM_017030148.1 | XP_016885637.1 | 1 | 477 | |||
XM_017030149.1 | XP_016885638.1 | 1 | 477 | |||
XM_017030150.1 | XP_016885639.1 | 1 | 477 | |||
XM_017030151.1 | XP_016885640.1 | 1 | 477 | |||
XM_017030152.1 | XP_016885641.1 | 1 | 477 | |||
XM_017030153.1 | XP_016885642.1 | 1 | 477 | |||
XM_017030154.1 | XP_016885643.1 | 1 | 477 | |||
XM_017030155.1 | XP_016885644.1 | 1 | 477 | |||
XM_017030156.1 | XP_016885645.1 | 1 | 477 | |||
XM_017030157.1 | XP_016885646.1 | 1 | 477 | |||
XM_017030158.1 | XP_016885647.1 | 1 | 477 | |||
XM_017030159.1 | XP_016885648.1 | 1 | 477 | |||
XM_017030160.1 | XP_016885649.1 | 1 | 477 | |||
XM_017030161.1 | XP_016885650.1 | 1 | 477 |
Gene Symbol: FAM230A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136560.1 | 4 | 1869 |
Gene Symbol: FAM230B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108107.1 | 9 | 2272 | |||
GenBank mRNA | AK128837.1 | - | 3 | 2086 | ||
BC039313.1 | - | 11 | 2165 |
Gene Symbol: LOC100996335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136559.1 | 8 | 2057 |
Gene Symbol: LOC100996415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136570.1 | 8 | 1883 |
Gene Symbol: LOC100996432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136571.1 | 8 | 2136 |
Gene Symbol: LOC105379550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136574.1 | 1 | 538 |