Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206836.1 | NP_001193765.1 | 2 | 297 | ||
NM_004663.4 | NP_004654.1 | 2 | 297 | |||
GenBank mRNA | AF000231.1 | - | 2 | 237 | ||
AF498946.1 | - | 2 | 170 | |||
AK297549.1 | - | 2 | 308 | |||
AK300008.1 | - | 2 | 215 | |||
AK311770.1 | - | 2 | 281 | |||
BC013348.2 | - | 2 | 264 | 3510339 | ||
BI550703.1 | - | 2 | 303 | |||
BT020151.1 | - | 2 | 170 | |||
BT020154.1 | - | 2 | 170 | |||
CR407669.1 | - | 2 | 170 | |||
CR536493.1 | - | 2 | 170 | |||
DC300743.1 | - | 2 | 307 | |||
X53143.1 | - | 2 | 170 | |||
X56740.1 | - | 2 | 223 |