Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205859.1 | NP_995581.1 | 1 | 280 | ||
XM_011518520.2 | XP_011516822.1 | 2 | 448 | |||
GenBank mRNA | BC111086.1 | - | 1 | 280 | 40023037 |