Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039845.2 | NP_001034934.1 | 8 | 1523 | ||
NM_001282940.1 | NP_001269869.1 | 8 | 1523 | |||
NR_104261.1 | 6 | 884 | ||||
XM_005246315.4 | XP_005246372.1 | 7 | 1344 | |||
XM_005246316.4 | XP_005246373.1 | 6 | 675 | |||
XM_011510628.2 | XP_011508930.1 | 8 | 1509 | |||
XM_017003373.1 | XP_016858862.1 | 7 | 1192 | |||
XM_017003374.1 | XP_016858863.1 | 7 | 1179 | |||
GenBank mRNA | AK058070.1 | - | 7 | 1153 | ||
AK292378.1 | - | 8 | 1521 | |||
AK292386.1 | - | 8 | 1260 | |||
AK302250.1 | - | 6 | 628 | |||
AK308257.1 | - | 8 | 1300 | |||
AK309144.1 | - | 7 | 1187 | |||
BC033509.1 | - | 6 | 629 | 5174046 | ||
BC088356.1 | - | 8 | 1277 | 30383553 |