Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004691.4 | NP_004682.2 | 1, 2 | 237 | ||
GenBank mRNA | AK300570.1 | - | 1, 2 | 169 | ||
AK313035.1 | - | 1, 2 | 195 | |||
BC008861.2 | - | 1, 2 | 215 | 4126712 | ||
BI552108.1 | - | 1, 2 | 251 |