Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207336.1 | NP_997219.1 | 5 | 488 | ||
XM_005249959.4 | XP_005250016.1 | 5 | 481 | |||
XM_005249960.4 | XP_005250017.1 | 5 | 410 | |||
XM_005249961.4 | XP_005250018.1 | 5 | 460 | |||
XM_006715864.3 | XP_006715927.1 | 6 | 590 | |||
XM_011515855.2 | XP_011514157.1 | 5 | 2060 | |||
XM_011515856.2 | XP_011514158.1 | 6 | 1763 | |||
XM_011515857.2 | XP_011514159.1 | 4 | 1950 | |||
XM_011515858.1 | XP_011514160.1 | 6 | 675 | |||
XM_017011799.1 | XP_016867288.1 | 3 | 1834 | |||
GenBank mRNA | BC029296.1 | - | 3 | 297 | ||
BC052625.1 | - | 5 | 488 | 6570263 |