Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207341.3 | NP_997224.2 | 5 | 987 | ||
XM_011544852.2 | XP_011543154.1 | 2 | 503 | |||
XM_011544853.2 | XP_011543155.1 | 3 | 366 | |||
GenBank mRNA | BC067899.1 | - | 1 | 586 | 5298273 | |
CR741030.1 | - | 1 | 257 | |||
EU794684.1 | - | 5 | 987 |