Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207341.3 | NP_997224.2 | 8 | 1275 | ||
XM_011544852.2 | XP_011543154.1 | 5 | 791 | |||
XM_011544853.2 | XP_011543155.1 | 6 | 654 | |||
GenBank mRNA | BC067899.1 | - | 4 | 874 | 5298273 | |
EU794684.1 | - | 8 | 1275 |