Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207352.3 | NP_997235.3 | 10 | 1567 | ||
XM_005262935.3 | XP_005262992.1 | 10 | 1349 | |||
XM_017008037.1 | XP_016863526.1 | 9 | 1325 | |||
GenBank mRNA | AK022114.1 | - | 4 | 440 | ||
AK122600.1 | - | 10 | 1302 | |||
AK126473.1 | - | 11 | 1236 | |||
AK311390.1 | - | 4 | 353 | |||
AY422002.1 | - | 10 | 1567 | |||
BC041839.1 | - | 3 | 1001 | |||
BC060857.1 | - | 10 | 1433 | 30333559 | ||
BX648537.1 | - | 4 | 498 | |||
BX648730.1 | - | 5 | 5321 |