Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145719.1 | NP_001139191.1 | 7 | 1198 | ||
NM_207372.2 | NP_997255.2 | 7 | 1694 | |||
XM_017016236.1 | XP_016871725.1 | 7 | 4112 | |||
GenBank mRNA | AK091518.1 | - | 2 | 339 | ||
AK123885.1 | - | 7 | 1694 | |||
AK123978.1 | - | 8 | 1198 |